missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse MYDGF Protein
A cDNA sequence encoding the MYDGF was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12766-5ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
MYDGF Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSVSEPTTV PFDVRPGGVV HSFSQDVGPG NKFTCTFTYA SQGGTNEQWQ MSLGTSEDSQ HFTCTIWRPQ GKSYLYFTQF KAELRGAEIE YAMAYSKAAF ERESDVPLKS EEFEVTKTAV SHRPGAFKAE LSKLVIVAKA ARSEL | |
Greater than 95.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
MYDGF protein solution (0.5 mg/ml) containing 20mM Tris-HCl (pH 8.0), 0.1M NaCl and 10% glycerol. |