missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Nuclease-sensitive element-binding protein 1
A DNA sequence encoding the Mus musculus (Mouse) Nuclease-sensitive element-binding protein 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9471-ye-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_035862.2 | |
Nuclease-sensitive element-binding protein 1 | |
Research Use Only | |
Ybx1 | |
Recombinant Protein | |
Yeast | |
SSEAETQQPP AAPAAALSAA DTKPGSTGSG AGSGGPGGLT SAAPAGGDKK VIATKVLGTV KWFNVRNGYG FINRNDTKED VFVHQTAIKK NNPRKYLRSV GDGETVEFDV VEGEKGAEAA NVTGPGGVPV QGSKYAADRN HYRRYPRRRG PPRNYQQNYQ NSESGEKNEG SESAPEGQAQ QRRPYRRRRF PPYYMRRPYA RRPQYSNPPV QGEVMEGADN QGAGEQGRPV RQNMYRGYRP RFRRGPPRQR QPREDGNEED KENQGDETQG QQPPQRRYRR NFNYRRRRPE NPKPQDGKET KAADPPAENS SAPEAEQGGA E | |
Greater than 90% as determined by SDS-PAGE. |
22608 | |
10 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |