missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Protein cereblon Protein
A DNA sequence encoding the Mus musculus (Mouse) Protein cereblon, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9830-ye-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_067424.2 | |
Protein cereblon Protein | |
Research Use Only | |
Crbn | |
Recombinant Protein | |
Yeast | |
MAGEGDQQDA AHNMGNHLPL LPADSEDEDD EIEMEVEDQD SKEARKPNII NFDTSLPTSH TYLGADMEEF HGRTLHDDDS CQVIPVLPEV LMILIPGQTL PLQLSHPQEV SMVRNLIQKD RTFAVLAYSN VQEREAQFGT TAEIYAYREE QEFGIEVVKV KAIGRQRFKV LELRTQSDGI QQAKVQILPE CVLPSTMSAV QLESLNKCQV FPSKPISWED QYSCKWWQKY QKRKFHCANL TSWPRWLYSL YDAETLMDRI KKQLREWDEN LKDDSLPENP IDFSYRVAAC LPIDDVLRIQ LLKIGSAIQR LRCELDIMNK CTSLCCKQCQ ETEITTKNEI FSLSLCGPMA AYVNPHGYVH ETLTVYKASN LNLIGRPSTV HSWFPGYAWT IAQCKICASH IGWKFTATKK DMSPQKFWGL TRSALLPTIP ETEDEISPDK VILCL | |
Greater than 80% as determined by SDS-PAGE. |
58799 | |
1 mg | |
Not determined. | |
Full. Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |