missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Retnla Protein
Recombinant Protein
Marca: enQuireBio™ QP5432-5ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_065255.2 | |
Retnla Protein | |
Research Use Only | |
Retnla | |
Mouse | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |
57262 | |
5 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MDETIEIIVE NKVKELLANP ANYPSTVTKT LSCTSVKTMN RWASCPAGMT ATGCACGFAC GSWEIQSGDT CNCLCLLVDW TTARCCQLS | |
Greater than 95% as determined by SDS-PAGE. |