missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Roundabout homolog 1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Roundabout homolog 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10099-ma-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_062286.2 | |
Roundabout homolog 1 Protein | |
Research Use Only | |
Robo1 | |
Recombinant Protein | |
Mammal | |
YRHRKKRNGL TSTYAGIRKV PSFTFTPTVT YQRGGEAVSS GGRPGLLNIS EPATQPWLAD TWPNTGNNHN DCSINCCTAG NGNSDSNLTT YSRPADCIAN YNNQLDNKQT NLMLPESTVY GDVDLSNKIN EMKTFNSPNL KDGRFVNPSG QPTPYATTQL IQANLSNNMN NGAGDSSEKH WKPPGQQKPE VAPIQYNIME QNKLNKDYRA NDTIPPTIPY NQSYDQNTGG SYNSSDRGSS TSGSQGHKKG ARTPKAPKQG GMNWADLLPP PPAHPPPHSN SEEYNMSVDE SYDQEMPCPV PPAPMYLQQD ELQEEEDERG PTPPVRGAAS SPAAVSYSHQ STATLTPSPQ EELQPMLQDC PEDLGHMPHP PDRRRQPVSP PPPPRPISPP HTYGYISGPL VSDMDTDAPE EEEDEADMEV AKMQTRRLLL RGLEQTPASS VGDLESSVTG SMINGWGSAS EEDNISSGRS SVSSSDGSFF TDADFAMS-Q AVAAAAEYAG LKVARRQMQD AAGRRHFHAS QCPRPTSPVS TDSNMSAVVI QKARPAKKQK HQPGHLRREA YADDLPPPPV PPPAIKSPTV QSKAQLEVRP VMVPKLASIE ARTDRSSDRK GGSYKGREAL DGRQVTDLRT NPSDPREAQE QPNDGKGRGT RQPKRDLPPA KTHLGQEDIL PYCRPTFPTS NNPRDPSSSS SMSSRGSGSR QREQANVGRR NMAEMQVLGG FERGDENNEE LEETES | |
Greater than 90% as determined by SDS-PAGE. |
19876 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |