missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse S100b Protein
A cDNA sequence encoding the S100b was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13403-5ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
S100b Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MSELEKAMVA LIDVFHQYSG REGDKHKLKK SELKELINNE LSHFLEEIKE QEVVDKVMET LDEDGDGECD FQEFMAFVAM VTTACHEFFE HE | |
Greater than 95.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
Untagged | |
The s100b protein solution (1 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |