missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mycoplasma 46 kDa surface antigen Protein
A DNA sequence encoding the Mycoplasma hyopneumoniae (strain 232) 46 kDa surface antigen, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7000-ye-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
WP_011206345.1 | |
10 μg | |
Not determined. | |
Full Length | |
Mycoplasma | |
His | |
Tris-based buffer, 50% glycerol |
Mycoplasma 46 kDa surface antigen Protein | |
Research Use Only | |
p46 | |
Recombinant Protein | |
Yeast | |
CGQTESGSTS DSKPQAETLK HKVSNDSIRI ALTDPDNPRW ISAQKDIISY VDETEAATST ITKNQDAQNN WLTQQANLSP APKGFIIAPE NGSGVGTAVN TIADKGIPIV AYDRLITGSD KYDWYVSFDN EKVGELQGLS LAAGLLGKED GAFDSIDQMN EYLKSHMPQE TISFYTIAGS QDDNNSQYFY NGAMKVLKEL MKNSQNKIID LSPEGENAVY VPGWNYGTAG QRIQSFLTIN KDPAGGNKIK AVGSKPASIF KGFLAPNDGM AEQAITKLKL EGFDTQKIFV TGQDYNDKAK TFIKDGDQNM TIYKPDKVLG KVAVEVLRVL IAKKNKASRS EVENELKAKL PNISFKYDNQ TYKVQGKNIN TILVSPVIVT KANVDNPDA | |
Greater than 86.9% as determined by SDS-PAGE. |