missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mycoplasma Methionine aminopeptidase Protein
A DNA sequence encoding the Mycoplasma pneumoniae (strain ATCC 29342 / M129) Methionine aminopeptidase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7522-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_109874.1 | |
Mycoplasma Methionine aminopeptidase Protein | |
Research Use Only | |
map | |
Recombinant Protein | |
E. coli | |
MVYLKSAREV EQIRQACKIF QEAKAYFTIE RLLGKSLTAI DQALKQFIES KGATCAFHKY QNFPGFNCLS LNETVIHGIA DNRVFGVKDK LTLDIGINLN GYICDAAFTV LGPKAPEPMQ TLLEVTEACF TAVVEPQLRP NNPTGNVSHA IQTYFESKGY YLLKQFGGHG CGIKVHEEPL ILNYGKPDTG TKLEPGMVLC IEPMVMTDSD AMVMHNNSWN VLTPKSRYNC HVEQMYVITT SGFECLTN | |
Greater than 90% as determined by SDS-PAGE. |
876883 | |
10 μg | |
Not determined. | |
Full Length | |
Mycoplasma | |
His-SUMO | |
Tris-based buffer, 50% glycerol |