Learn More
enQuireBio™ Recombinant other B. Microti IRA Protein
A cDNA sequence encoding the B. Microti IRA was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11115-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
B. Microti IRA Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
B. Microti IRA is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
Greater than 80.0% as determined by SDS-PAGE. |