missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Batroxobin Protein
A cDNA sequence encoding the Batroxobin was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10529-2ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
Batroxobin Protein | |
Research Use Only | |
Batroxobin’s biological activity was found to be of no less than 500KU/mg. (Klobusitzky Unit). One KU is defined as the amount of enzyme which coagulates standard human plasma incubated at 37C in vitro within 60 ± 20 seconds. | |
Other | |
Untagged | |
The Batroxobin protein was lyophilized from a concentrated (1 mg/ml) solution containing 20mM sodium acetate buffer, pH 7.4. |
2 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
P. pastoris | |
VIGGDECDIN EHPFLAFMYY SPRYFCGMTL INQEWVLTAA HCNRRFMRIH LGKHAGSVAN YDEVVRYPKE KFICPNKKKN VITDKDIMLI RLDRPVKNSE HIAPLSLPSN PPSVGSVCRI MGWGAITTSE DTYPDVPHCA NINLFNNTVC REAYNGLPAK TLCAGVLQGG IDTCGGDSGG PLICNGQFQG ILSWGSDPCA EPRKPAFYTK VFDYLPWIQS IIAGNKTATC P | |
Greater than 97.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |