missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Batroxobin Protein

A cDNA sequence encoding the Batroxobin was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP10529-10ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15954148

  • 212.00€ / 10µg
Spedizione stimata: 28-05-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Specifica

Specifica

Batroxobin Protein
Research Use Only
Batroxobin’s biological activity was found to be of no less than 500KU/mg. (Klobusitzky Unit). One KU is defined as the amount of enzyme which coagulates standard human plasma incubated at 37C in vitro within 60 ± 20 seconds.
Other
Untagged
The Batroxobin protein was lyophilized from a concentrated (1 mg/ml) solution containing 20mM sodium acetate buffer, pH 7.4.
10 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
P. pastoris
VIGGDECDIN EHPFLAFMYY SPRYFCGMTL INQEWVLTAA HCNRRFMRIH LGKHAGSVAN YDEVVRYPKE KFICPNKKKN VITDKDIMLI RLDRPVKNSE HIAPLSLPSN PPSVGSVCRI MGWGAITTSE DTYPDVPHCA NINLFNNTVC REAYNGLPAK TLCAGVLQGG IDTCGGDSGG PLICNGQFQG ILSWGSDPCA EPRKPAFYTK VFDYLPWIQS IIAGNKTATC P
Greater than 97.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali