missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other DsbC Protein
A cDNA sequence encoding the DsbC was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11704-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
947363 | |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
DDAAIQQTLA KMGIKSSDIQ PAPVAGMKTV LTNSGVLYIT DDGKHIIQGP MYDVSGTAPVNVTNKMLLKQ LNALEKEMIV YKAPQEKHVI VFTDITCGY CHKLHEQMAD YNALGITVRYLAFPRQGLDS DAEKEMKAIW CAKDKNKAFD DVMAGKSVAP ASCDVDIADH YVLGVQLGVSGTPAVVLSNG TLVPGYQPPK EMKEFLDEHQ KMTSGK | |
Greater than 95.0% as determined by SDS-PAGE. |
DsbC Protein | |
Research Use Only | |
dsbC | |
Other | |
Untagged | |
1 mg/ml solution containing 20mM phosphate buffer pH 7.5 and 20mM EDTA. |