missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other DUT Pyrococcus Fruriosus Protein
A cDNA sequence encoding the DUT Pyrococcus Fruriosus was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10580-500IU
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
DUT Pyrococcus Fruriosus Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
dUTPase is supplied in 20mM Tris-HCl (pH 8.2), 1mM DTT, 0.1mM EDTA, 100mM KCl, 0.1% Nonidet P40, 0.1% Tween 20 and 50% glycerol at a concentration of 10U/microL of the enzyme. |
500 IU | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Other | |
Untagged | |
Greater than 97.0% as determined by SDS-PAGE. |