Learn More
enQuireBio™ Recombinant other Elcatonin Protein
Recombinant Protein
Marca: enQuireBio™ QP10585-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
Elcatonin Protein | |
Research Use Only | |
Biological Activity (based on net peptide) was found to be 6695.2 IU/mg. | |
Other | |
Ser-Asn-Leu-Ser-Thr-Asu-Val-Leu-Gly-Lys-Leu-Ser-Gln-Glu-Leu-His-Lys-Leu-Gln-Thr-Tyr-Pro-Arg-Thr-Asn-Val-Gly-Ala-Gly-Thr-Pro-NH2 | |
Greater than 93.3% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Untagged | |
The protein was lyophilized with no additives. |