missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other scIC Protein
A cDNA sequence encoding the scIC was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13432-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
scIC Protein | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged |
Greater than 85.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Each mg/ml of protein contains 50% glycerol. |