missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other SecB Protein
A cDNA sequence encoding the SecB was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13448-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
SecB Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MSEQNNTEMT FQIQRIYTKD ISFEAPNAPH VFQKDWQPEV KLDLDTASSQ LADDVYEVVL RVTVTASLGE ETAFLCEVQQ GGIFSIAGIE GTQMAHCLGA YCPNILFPYA RECITSMVSR GTFPQLNLAP VNFDALFMNY LQQQAGEGTE EHQDA | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
The SecB protein solution (1 mg/ml) contains 20mM Tris-HCl pH 8 and 10% glycerol. |