missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Thymosin ?4 Protein

A cDNA sequence encoding the Thymosin ?4 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13746-20ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15944569

  • 120.00€ / 20µg

Spedizione stimata: 31-01-2023
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

Specifica

Specifica

Thymosin ?4 Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
Lyophilized from a 0.2um filtered concentrated solution in 1xPBS, pH 7.4.
Greater than 95.0% as determined by SDS-PAGE and HPLC analyses.
Research Use Only
Recombinant Protein
E. coli
SDKPDMAEIE KFDKSKLKKT ETQEKNPLPS KETIEQEKQA GES
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali