missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant P. gingivalis Gingipain R1 Protein
A DNA sequence encoding the Porphyromonas gingivalis Gingipain R1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7192-ye-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
P. gingivalis Gingipain R1 Protein | |
Research Use Only | |
rgpA | |
Recombinant Protein | |
Yeast | |
YTPVEEKQNG RMIVIVAKKY EGDIKDFVDW KNQRGLRTEV KVAEDIASPV TANAIQQFVK QEYEKEGNDL TYVLLVGDHK DIPAKITPGI KSDQVYGQIV GNDHYNEVFI GRFSCESKED LKTQIDRTIH YERNITTEDK WLGQALCIAS AEGGPSADNG ESDIQHENVI ANLLTQYGYT KIIKCYDPGV TPKNIIDAFN GGISLVNYTG HGSETAWGTS HFGTTHVKQL TNSNQLPFIF DVACVNGDFL FSMPCFAEAL MRAQKDGKPT GTVAIIASTI NQSWASPMRG QDEMNEILCE KHPNNIKRTF GGVTMNGMFA MVEKYKKDGE KMLDTWTVFG DPSLLVRTLV PTKMQVTAPA QINLTDASVN VSCDYNGAIA TISANGKMFG SAVVENGTAT INLTGLTNES TLTLTVVGYN KETVIKTINT NGEPNPYQPV SNLTATTQGQ KVTLKWDAPS TKTNATTNTA RSVDGIRELV LLSVSDAPEL LRS | |
Greater than 80% as determined by SDS-PAGE. |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
P. gingivalis | |
His | |
Tris-based buffer, 50% glycerol |