missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Pig Vasopressin V2 receptor Protein
A DNA sequence encoding the Sus scrofa (Pig) Vasopressin V2 receptor, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5701-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_999397.1 | |
Vasopressin V2 receptor Protein | |
Research Use Only | |
AVPR2 | |
Recombinant Protein | |
E. coli | |
WSVWDPKAPR EGPPFVLLML LASLNSCTNP WIYASFSSSI SSELRSLLCC PRRRTPPSLR PQEESCATAS SFSARDTSS | |
Greater than 90% as determined by SDS-PAGE. |
397462 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Pig | |
His-SUMO | |
Tris-based buffer, 50% glycerol |