missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Porcine Acrp30 Protein

A cDNA sequence encoding the Acrp30 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP10947-10ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15945658

  • 181.00€ / 10µg

Spedizione stimata: 07-02-2023
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

Specifica

Specifica

Acrp30 Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Porcine
Flag Tag
Sterile filtered and lyophilized from 0.5 mg/ml in 20mM Tris buffer and 50mM NaCl pH 7.5.
Greater than 90% as determined by SDS PAGE.
Research Use Only
Recombinant Protein
HEK 293
HVDYKDDDDK PAGETTEKPG ALLPMPKGAC AGWMAGIPGH PGHNGTPGRD GRDGVPGEKG EKGDTGLTGP KGDTGESGVT GVEGPRGFPG IPGRKGEPGE SAYVYRSAFS VGLETRVTVP NMPIRFTKIF YNQQNHYDVT TGKFHCNIPG LYYFSFHITV LKDVKVSLYK DKAVLFTYDQ QDKNVDQASG VLLYLEKGDQ WLQAYGDEEN GVYADNVNDS FTGFLLYHNIE
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali