missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Porcine Procalcitonin Protein
A cDNA sequence encoding the Procalcitonin was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13153-2ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
Procalcitonin Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHASTPLRSALETL PDPGPLSEKE GRLLLAALVK AYVQRKTNEL EQEQEQETEG SSLDSSRAKR CSNLSTCVLS AYWRNLNNFH RFSGMGFGPE TPGKKSDIAS SLERDLFPRG MPQDAN | |
Greater than 95.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Porcine | |
His | |
Procalcitonin was filtered (0.4microm) and lyophilized in 20mM TRIS and 50mM NaCl, pH 8.0. |