missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rabbit GH Protein
A cDNA sequence encoding the GH was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11963-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
GH Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MAAGSWTAGL LAFALLCLPW PQEASAFPAM PLSSLFANAV LRAQHLHQLA ADTYKEFERA YIPEGQRYSI QNAQAAFCFS ETIPAPTGKD EAQQRSDMEL LRFSLLLIQS WLGPVQFLSR AFTNTLVFGT SDRVYEKLKD LEEGIQALMR ELEDGSPRVG QLLKQTYDKF DTNLRGDDAL LKNYGLLSCF KKDLHKAETY LRVMKCRRFV ESSCVF | |
Greater than 97.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Rabbit | |
Untagged | |
The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.0045mM NaHCO3. |