missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat GDNF Protein
Recombinant Protein
Marca: enQuireBio™ QP5503-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_062012.1 | |
GDNF Protein | |
Research Use Only | |
GDNF | |
Recombinant Protein | |
E. coli | |
MSPDKQAAAL PRRERNRQAA AASPENSRGK GRRGQRGKNR GCVLTAIHLN VTDLGLGYET KEELIFRYCS GSCEAAETMY DKILKNLSRS RRLTSDKVGQ ACCRPVAFDD DLSFLDDSLV YHILRKHSAK RCGCI | |
Greater than 95% as determined by SDS-PAGE. |
25453 | |
500 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Rat GDNF induces C6 cell proliferation (typical ED50 is ?0.1 ug/ml). | |
Rat | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |