missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat IL-1 beta / IL1B Protein
Recombinant Protein
Marca: enQuireBio™ QP5516-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
IL-1 beta / IL1B Protein | |
Research Use Only | |
IL1B | |
Recombinant Protein | |
E. coli | |
MVPIRQLHCR LRDEQQKCLV LSDPCELKAL HLNGQNISQQ VVFSMSFVQG ETSNDKIPVA LGLKGLNLYL SCVMKDGTPT LQLESVDPKQ YPKKKMEKRF VFNKIEVKTK VEFESAQFPN WYISTSQAEH RPVFLGNSNG RDIVDFTMEP VSS | |
Greater than 95% as determined by SDS-PAGE. |
10 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Rat IL-1 beta / IL1B induces D10S cell proliferation (typical ED50 is ? 25 ng/ml). | |
Rat | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |