missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat IL25 Protein
Recombinant Protein
Marca: enQuireBio™ QP5512-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001178936.1 | |
IL25 Protein | |
Research Use Only | |
IL25 | |
Recombinant Protein | |
E. coli | |
MCSHLPRCCP SKQQEFPEEW LKWNPAPVSP PEPLRHTHHP ESCRASKDGP LNSRAISPWS YELDRDLNRV PQDLYHARCL CPHCVSLQTG SHMDPMGNSV PLYHNQTVFY RRPCHGEQGA HGRYCLERRL YRVSLACVCV RPRMMA | |
Greater than 95% as determined by SDS-PAGE. |
501996 | |
10 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Rat IL25 induces IL-8 production from human PBMCs at concentrations greater than or equal to 100 ng/ml. | |
Rat | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |