missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat RAP Protein
A cDNA sequence encoding the RAP was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13264-5ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
RAP Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MAPLRDRVSTLPRLQLLVLLLLPLLLVPQPIAGHGGKYSREKNEPEMAAKRESGEEFRME KLNQLWEKAKRLHLSPVRLAELHSDLKIQERDELNWKKLKVEGLDGDGEKEAKLVHNLNV ILARYGLDGRKDTQTVHSNALNEDTQDELGDPRLEKLWHKAKTSGKFSSEELDKLWREFL HYKEKIHEYNVLLDTLSRAEEGYENLLSPSDMTHIKSDTLASKHSELKDRLRSINQGLDR LRKVSHQGYGPATEFEEPRVIDLWDLAQSANFTEKELESFREELKHFEAKIEKHNHYQKQ LEISHQKLKHVESIGDPEHISRNKEKYVLLEEKTKELGYKVKKHLQDLSSRVSRARHNEL | |
Greater than 95.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Rat | |
His | |
The protein (1 mg/ml) was lyophilized after from a sterile solution containing TBS pH 7.5, 0.1% BSA and 0.09% NaN3. |