missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat REG3B / Pap1 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) REG3B / Pap1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7120-200ug
Additional Details : Peso : 0.01000kg
Specifications
NP_445741.1 | |
REG3B / Pap1 Protein | |
Research Use Only | |
Reg3b | |
Recombinant Protein | |
E. coli | |
EDSPKKIPSA RISCPKGSQA YGSYCYALFQ IPQTWFDAEL ACQKRPEGHL VSVLNVAEAS FLASMVKNTG NSYQYTWIGL HDPTLGGEPN GGGWEWSNND IMNYVNWERN PSTALDRGFC GSLSRSSGFL RWRDTTCEVK LPYVCKFTG | |
Greater than 90% as determined by SDS-PAGE. |
24618 | |
200 μg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |