missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat S100G Protein
A cDNA sequence encoding the S100G was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13406-1ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
24249 | |
1 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MSAKKSPEEM KSIFQKYAAK EGDPNQLSKE ELKLLIQSEF PSLLKASSTL DNLFKELDKN GDGEVSYEEF EVFFKKLSQ | |
Greater than 90% as determined by SDS-PAGE. |
S100G Protein | |
Research Use Only | |
S100g | |
Rat | |
His | |
The protein was lyophilized from a concentrated solution (1 mg/ml) containing 100mM Phosphate buffer, pH 7.3. |