missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat S100G Protein

A cDNA sequence encoding the S100G was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13406-5ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15953469

  • 181.00€ / 5µg

Spedizione stimata: 03-02-2023
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

Specifica

Specifica

24249
Greater than 90% as determined by SDS-PAGE.
Research Use Only
S100g
Rat
His
The protein was lyophilized from a concentrated solution (1 mg/ml) containing 100mM Phosphate buffer, pH 7.3.
S100G Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MSAKKSPEEM KSIFQKYAAK EGDPNQLSKE ELKLLIQSEF PSLLKASSTL DNLFKELDKN GDGEVSYEEF EVFFKKLSQ
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali