missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus Replication initiation Protein
A DNA sequence encoding the Staphylococcus aureus Replication initiation protein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7349-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_052693.1 | |
S. aureus Replication initiation Protein | |
Research Use Only | |
repD | |
Recombinant Protein | |
E. coli | |
MSTENHSNYL QNKDLDNFSK TGYSNSRLSG NFFTTPQPEL SFDAMTIVGN LNKTNAKKLS DFMSTEPQIR LWDILQTKFK AKALQEKVYI EYDKVKADSW DRRNMRVEFN PNKLTHEEML WLKQNIIDYM EDDGFTRLDL AFDFEDDLSD YYAMTDKAVK KTIFYGRNGK PETKYFGVRD SDRFIRIYNK KQERKDNADV EVMSEHLWRV EIELKRDMVD YWNDCFDDLH ILKPDWTTPE KVKEQAMVYL LLNEEGTWGK LERHAKYKYK QLIKEISPID LTELMKSTLK ENEKQLQKQI DFWQREFRFW K | |
Greater than 90% as determined by SDS-PAGE. |
3355242 | |
50 μg | |
Not determined. | |
Full Length | |
S. aureus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |