missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant SARS SARS Mosaic S(C) Protein

A cDNA sequence encoding the SARS Mosaic S(C) was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13420-1mg

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15953509

  • 1330.00€ / 1mg

Spedizione stimata: 07-02-2023
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

Specifica

Specifica

SARS SARS Mosaic S(C) Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
SARS
Untagged
SARS Mosaic Protein is >95% pure as determined by 10% PAGE (coomassie staining).
Research Use Only
Recombinant Protein
E. coli
25mM Tris-HCl, 0.4% sarcosyl, 0.25% Triton .–100 and 50% glycerol.
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali