Learn More
Corning™ Step-R™ Repeating Pipettor Syringe Tips, Sterile
Corning™ Step-R™ Repeating Pipettor Syringe Tips, Sterile provide precise, positive-displacement dispensing of viscous or volatile liquids.
Marca: Corning™ 6633
Dettagli aggiuntivi : Peso : 5.00000kg
Descrizione
- Ideal for viscous, dense and volatile liquides
- Minimum increments printed on the syringe itself
- Accurately deliver selected dose
- Made from high quality polypropylene and polyethylene
- Pyrogen-, ATP-, RNase-, and DNase-free
Specifica
Autoclavable | |
Polypropylene | |
Dispensing of viscous and volatile liquids | |
12.5 mL |
ISO8655 ISO11137 | |
Sterile | |
12.5 mL |