missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RFC4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10806-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Human Replication Factor C (RFC) is a hetero-pentameric protein consisting of the p140, p40, p38, p37 and p36 subunits. RFC functions in the loading of the DNA polymerase delta processivity factor (PCNA, Proliferating Cell nuclear Antigen) onto a primed DNA template during DNA synthesis and repair. RFC may also participate in the DNA damage checkpoint response with a PCNA-like hetero-trimer consisting of Hus 1, Rad 1 and Rad 9.Specifica
RFC4 | |
Polyclonal | |
Unconjugated | |
RFC4 | |
A1, A1 37 kDa subunit, Activator 1 37 kDa subunit, Activator 1 subunit 4, Replication factor C subunit 4, RF C 37 kDa subunit, RFC37, RFC4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5984 | |
-20°C | |
Liquid |
Immunofluorescence, Flow Cytometry, Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P35249 | |
RFC4 | |
RFC4 Fusion Protein Ag1234 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |