missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RXRG Mouse anti-Human, Clone: 1E7E5, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66306-1-IG-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Retinoic Acid (RA; active metabolite of vitamin A) plays a prominent role in regulating the transition of proliferating precursor cells (such as carcinoma cells and neuronal precursors) to postmitotic differentiated cells (Joshi et al., 2005). The Retinoid X Receptors (RXRs) family (RXR alpha, beta, and gamma), preferentially bind 9-cis -RA and regulate gene transcription by forming heterodimers with a second family of RA receptors. RAs have been suggested to potentially play a therapeutic role in cervical cancer (Abu et al., 2005). RAs are known to play key roles in neuronal development and an increasing body of evidence indicates that retinoid signaling may regulate synaptic plasticity and associated learning and memory behaviors (Lane and Bailey, 2005).Specifica
RXRG | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P48443 | |
RXRG | |
RXRG Fusion Protein Ag1611 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Western Blot | |
1E7E5 | |
Unconjugated | |
RXRG | |
NR2B3, Retinoid X receptor gamma, retinoid X receptor, gamma, RXRC, RXRG | |
Mouse | |
Protein A | |
RUO | |
6258 | |
-20°C | |
Liquid |