Learn More
ACROBiosystems SARS-CoV-2 S1 protein NTD (L18F, D80A, D215G, 242-244del, R246I), His Tag
Marca: ACROBiosystems S1D-C52Hc-1mg
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 34.3 kDa.Specifica
34.3 kDa | |
SARS-CoV-2 S1 protein NTD, His Tag (S1D-C52Hc) is expressed from human 293 cells (HEK293). It contains AA Ser 13 - Leu 303 (Accession # QHD43416.1). | |
-20°C | |
S1 protein NTD | |
SARS-CoV-2 | |
HEK293 | |
Viral Proteins |
1 mg | |
Ser 13 - Leu 303 | |
S1 protein NTD,Spike protein S1 NTD,BetaCoV S1-NTD,COVID-19 | |
Unconjugated | |
His Tag | |
Powder |