Learn More
ACROBiosystems SARS-CoV-2 S1 protein NTD (L18F, D80A, D215G, 242-244del, R246I), His Tag
Marca:  ACROBiosystems S1D-C52Hc-100ug
Dettagli aggiuntivi : Peso : 0.00010kg
Codice prodotto. 17226450
  • 414.60€ / 100µg
Spedizione stimata: 11-04-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 34.3 kDa.
Specifica
Specifica
34.3 kDa
SARS-CoV-2 S1 protein NTD, His Tag (S1D-C52Hc) is expressed from human 293 cells (HEK293). It contains AA Ser 13 - Leu 303 (Accession # QHD43416.1).
-20°C
S1 protein NTD
SARS-CoV-2
HEK293
Viral Proteins
100 μg
Ser 13 - Leu 303
S1 protein NTD,Spike protein S1 NTD,BetaCoV S1-NTD,COVID-19
Unconjugated
His Tag
Powder