Learn More
Sartorius Quantitative Grade 292 Filter Papers
These filter papers are especially suitable for qualitative analyses that require a low ash content.
Marca: Sartorius FT-3-205-240
Dettagli aggiuntivi : Peso : 0.10000kg
Descrizione
- Pure cotton linters or cotton linters with refined pulp
- No additives, such as wet-strengthening agents
Specifica
Disc | |
60 sec. | |
100/Pk. | |
240 mm |
5 to 8 μm | |
292 | |
0.18 mm |