Learn More
SI Analytics™ ScienceLine™ pH Combination Electrode
SI Analytics™ ScienceLine™ pH Combination Electrode includes a plug head connection and a fixed cable.
Brand: SI Analytics™ 285101252
Additional Details : Peso : 0.80000kg
Description
- Reference system: Silamid™
- Glass shaft
- Zero point: pH = 7.0 ± 0.3
- Fixed Cable: 1 m long, with plug A acc. to DIN 19262 or with BNC plug
Specifications
pH glass A | |
0 to 14 | |
Sphere | |
120 mm |
Ceramic | |
pH Combination Electrode | |
-5°C to +80°C | |
12 mm |