Learn More
SDC4 Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca:  Proteintech 11820-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Codice prodotto. 16899143
  • 121.38€ / 20µL
Spedizione stimata: 11-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
SDC4, also named as syndecan 4, is a cell surface proteoglycan that bears heparan sulfate, which is involved in normal wound healing. It has been reported that SDC4 may be associated with keratinocyte proliferation and differentiation in the epidermis. SDC4 also plays a key role in the angiogenesis and inflammation. The predicted MW of SDC4 is 22 kDa and 11820-1-AP antibody recognizes a protein around 24-26 kDa and 48-52 kDa (homodimer) which is similar to the papers.(PMID: 20639201, 21949132; 18638267; 30165731 )
Specifica
Specifica
SDC4
Polyclonal
Unconjugated
20 μL
6385
Liquid
ELISA, Immunofluorescence, Immunohistochemistry, Western Blot
600 μg/mL
Rabbit
Primary
Mouse, Human