Learn More
Secretogranin V Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10761-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Acts as a molecular chaperone for PCSK2/PC2, preventing its premature activation in the regulated secretory pathway. Binds to inactive PCSK2 in the endoplasmic reticulum and facilitates its transport from there to later compartments of the secretory pathway where it is proteolytically matured and activated. Also required for cleavage of PCSK2 but does not appear to be involved in its folding. Plays a role in regulating pituitary hormone secretion. The C-terminal peptide inhibits PCSK2 in vitro.Specifica
Secretogranin V | |
Polyclonal | |
Unconjugated | |
SCG5 | |
7B2, Neuroendocrine protein 7B2, P7B2, Pituitary polypeptide, SCG5, Secretogranin 5, Secretogranin V, secretogranin V (7B2 protein), SGNE1, SgV | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20394, 25719, 6447 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P05408, P12961, P27682 | |
SCG5 | |
Secretogranin V Fusion Protein Ag1107 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |