missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEPT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11370-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Septin 1 (SEPT1) is a member of the septin family of GTPases. Members of this family are required for cytokinesis and the maintece of cellular morphology. This gene encodes a protein that can form homo- and heterooligomeric filaments, and may contribute to the formation of neurofibrillary tangles in Alzheimer′s disease. Alternatively spliced transcript variants have been found but the full-length nature of these variants has not been determined.Specifica
SEPT1 | |
Polyclonal | |
Unconjugated | |
SEPT1 | |
DIFF6, LARP, Peanut like protein 3, PNUTL3, SEP1, SEPT1, septin 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1731, 293507, 54204 | |
-20°C | |
Liquid |
Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P42209, Q5EB96, Q8WYJ6 | |
SEPT1 | |
SEPT1 Fusion Protein Ag1926 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |