Learn More
SGE™ Beckman/Altex, Rheodyne, SSI, Valco Valves with Fixed Needle
Used with Rheodyne™ and Valco™ Valves. SGE Analytical Science™ Beckman/Altex, Rheodyne, SSI, Valco Valves with Fixed Needle have a 22 gauge.
Marca: SGE™ 002335
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
- Fixed needle
- For use with For Rheodyne™ and Valco™ Valves
Specifica
Syringe | |
Beckman/Altex, Rheodyne™, SSI instruments, and valco valves | |
50.8 mm | |
0.71 mm |
22 G | |
Removable Needle Syringe with Removable Plunger | |
0.028 in. | |
10 μL |