Learn More
SGE™ SilFlow™ GC 3 Port Splitter: Device Only
Highly efficient and reliable micro fluidic platform to improve GC connectivity
Marca: SGE™ 123723
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
- 1.1mm OD tubing not included
Specifica
Port Splitter | |
1.1 mm | |
Ports B and C 0.25/0.32 mm | |
SGE Silflow Microchannel Device |