missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SGTA Mouse anti-Human, Mouse, Clone: 3B10B11, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 60305-1-IG-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This gene encodes a protein which is capable of interacting with the major nonstructural protein of parvovirus H-1 and 70-kDa heat shock cognate protein; however, its function is not known. Since this transcript is expressed ubiquitously in various tissues, this protein may serve a housekeeping function.Specifica
SGTA | |
Monoclonal | |
2.03 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O43765, Q8BJU0 | |
Sgta | |
SGTA Fusion Protein Ag1475 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG2b |
Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry | |
3B10B11 | |
Unconjugated | |
SGTA | |
Alpha SGT, alphaSGT, hSGT, SGT, SGT1, SGTA, UBP, Vpu binding protein | |
Mouse | |
Protein A | |
RUO | |
52551, 6449 | |
-20°C | |
Liquid |