Learn More
Silica 36-200μm, 60Å, EcoChrom™, MP Biomedicals™
For Chromatography; Chemical Synthesis
Marca: MP Biomedicals 0204667
Dettagli aggiuntivi : CAS : 7631-86-9 Peso : 25.00000kg
Specifica
25 kg | |
Silica | |
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
7631-86-9 |
Store at Room Temperature. | |
Powder | |
2g/cm3 at 20°C |