missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC38A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12039-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Amino acid transporters play essential roles in the uptake of nutrients, production of energy, chemical metabolism, detoxification, and neurotransmitter cycling. SLC38A1 is an important transporter of glutamine, an intermediate in the detoxification of ammonia and the production of urea. Glutamine serves as a precursor for the synaptic transmitter, glutamate.Specifica
SLC38A1 | |
Polyclonal | |
Unconjugated | |
SLC38A1 | |
Amino acid transporter A1, ATA1, NAT2, SAT1, SLC38A1, SNAT1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
105727, 170567, 81539 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8K2P7, Q9H2H9, Q9JM15 | |
Slc38a1 | |
SLC38A1 Fusion Protein Ag2668 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |