missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Smooth Surface Filter Discs Qualitative Filter Papers, Grade: 3 w

Used for analytical and technical purposes.

Marca:  Sartorius™ FT-3-308-110

Dettagli aggiuntivi : Peso : 0.10000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 10317841

  • 11.30€ / pezzi 100
Spedizione stimata: 10-06-2024
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

  • For routine analyses like clarification, determination of substances
  • Discs with a center hole for technical applications
  • Made of refined pulp and cotton linters with >95% α-cellulose content
  • Smooth surface
  • Wet-strengthened
Specifica

Specifica

Disc
15μm/sec.
3 w
0.14 mm
110 mm
Cotton Linters, Refined Pulp
For qualitative-technical analyses, used for routine analyses like clarification, determination of substances, but also as discs with a center hole for technical applications
100 Pack
Qualitative Filter Paper
110 mm
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali