Learn More
Spectrum Labs™ Spectra/Por 4 Dialysis Membrane Discs, 12-14kD MWCO
Spectrum Labs™ Spectra/Por 4 Dialysis Membrane Discs are for basic dialysis and are hydrophilic with symmetric porosity.
Marca: Spectrum Labs™ 132 498
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
- Dry Standard RC dialysis membrane in single-layer discs.
- Used in various commercially available dialysis equipment.
- Discs are separated by gylcine paper
- Packaged dry with glycerin (rinse out with water)
Specifica
Dialysis Membrane Discs | |
33 mm | |
Basic dialysis |
12 to 14 kDa | |
50/Pk. |