Learn More
Gilac™ Stainless Steel Trolley for Large Volume Container
Stainless Steel Trolley for Large Volume Container makes it easier to work at breast height. Equipped with 125 mm diameter wheels in white PA (2 fixed and 2 pivoting with brakes).
Marca:  Gilac™ G111595
Dettagli aggiuntivi : Peso : 8.80000kg
Codice prodotto. 17918661
  • 979.01€ / Pezzo
Spedizione stimata: 04-06-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
  • Trolley equiped with 125 mm diameter wheels in white PA (2 fixed and 2 pivoting with brakes).
  • Stainless steel recommended in humid environments (no damage to bearings even in the presence of water).
  • Makes it easier to work at breast height.
Specifica
Specifica
Trolley
Stainless Steel
170 L Large Volume Container
700 mm
Large Volume Containers
125 mm
27.6 in.