missing translation for 'onlineSavingsMsg'
Learn More

STEAP2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 20201-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16899714

  • 140.00€ / 20µL
Spedizione stimata: 15-05-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

The antibody is specific to STEAP2.

The six-transmembrane epithelial antigen of prostate 2 (STEAP2) is a member of a family of metalloreductases identified as cell-surface antigens in prostate tissue. Similar to two other members of the STEAP family (STEAP 3 and STEAP4), STEAP2 promotes both iron and copper reduction. STEAP2 expression in transfected cells also correlated with iron or copper uptake, suggesting that the STEAP family of proteins may function to stimulate iron and copper uptake. STEAP2 is widely expressed in many tissues in the plasma membrane, but is most highly expressed in prostate. At least three isoforms of STEAP2 are known to exist.
Specifica

Specifica

STEAP2
Polyclonal
Unconjugated
STEAP2
IPCA1, Metalloreductase STEAP2, PCANAP1, PUMPCn, STAMP1, STEAP2, STMP
Rabbit
Antigen Affinity Chromatography
RUO
261729, 74051
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.28 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8BWB6, Q8NFT2
Steap2
Peptide
20 μL
Primary
Mouse, Human
Antibody
IgG
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali